Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-ethoxyphenol | 622-62-8 | MFCD00002334 | 99.0% | 138.16 g/mol | C8H10O2 | 50 G
SDP

Catalog No. 5000301537
Encompass_Preferred

4-ethoxyphenol is an aromatic phenolic compound used as a building block, analytical standard, and research reagent. It is typically a white to off-white powder used in studies of antibacterial activity and in synthetic chemistry. The compound is supplied at high purity and should be handled according to safety data sheet guidance for phenolic substances.

  • High purity (commonly supplied at 99.0%)
  • Molecular weight 138.16 g/mol
  • Chemical formula C8H10O2
  • Appearance: white to off-white powder
  • Used as a building block and analytical standard in research
  • Handle according to safety data sheet recommendations

Catalog No. 50-003-01537 Supplier Medchemexpress LLC Supplier No. HYW01578650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-ethoxyphenol is an aromatic phenolic compound used as a building block, analytical standard, and research reagent. It is typically a white to off-white powder used in studies of antibacterial activity and in synthetic chemistry. The compound is supplied at high purity and should be handled according to safety data sheet guidance for phenolic substances.

  • High purity (commonly supplied at 99.0%)
  • Molecular weight 138.16 g/mol
  • Chemical formula C8H10O2
  • Appearance: white to off-white powder
  • Used as a building block and analytical standard in research
  • Handle according to safety data sheet recommendations

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.