Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxyphenyl isocyanate, 97%

Catalog No. AAL1150914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1150914 25 g
AAL1150906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1150914 Supplier Thermo Scientific Chemicals Supplier No. L1150914
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 32459-62-4 | C9H9NO2 | 163.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32459-62-4
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00013877
InChI Key FMYVTFRADSNGDN-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl isocyanate, benzene, 1-ethoxy-4-isocyanato, 4-ethoxyphenylisocyanate, 4-isocyanatophenetole, isocyanic acid 4-ethoxyphenyl ester, p-ethoxy phenyl isocyanate, 4-ethoxybenzenisocyanate, pubchem3178, p-athoxyphenylisocyanat, acmc-1afep
PubChem CID 122887
IUPAC Name 1-ethoxy-4-isocyanatobenzene
SMILES CCOC1=CC=C(C=C1)N=C=O

Specifications

Density 1.11
Boiling Point 235°C
Flash Point 105°C (221°F)
Refractive Index 1.536
Quantity 25 g
UN Number UN2206
Beilstein 742647
Sensitivity Moisture sensitive; May form a precipitate
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 4-Ethoxyphenyl isocyanate
Show More Show Less
Hazard Category H302+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.