Learn More
4-Ethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| Melting Point | 168°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Synonym | 4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic |
| SMILES | B(C1=CC=C(C=C1)OCC)(O)O |
| Molecular Weight (g/mol) | 165.983 |
| Formula Weight | 165.98 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Ethoxyphenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
| TSCA | No |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.