Learn More
Medchemexpress LLC 4-ethyl-1,3-benzenediol | 2896-60-8 | MFCD00002283 | 99.7% | 138.17 g/mol | C8H10O2 | 25 G
SDP
Catalog No. 5000304120
Encompass_Preferred
4-Ethylresorcinol is a research-grade aromatic diol used in biochemical and cosmetic research to study pigmentation and tyrosinase activity. It has reported antioxidant activity and can inhibit melanin synthesis, making it useful for depigmentation studies. The compound is supplied as a high-purity solid and is provided with supporting documentation for research use.
  • Derivative of resorcinol that serves as a substrate for tyrosinase.
  • Reported antioxidant properties and inhibition of melanin synthesis.
  • High purity (99.71%) suitable for analytical and research assays.
  • Supplied as a solid powder with available pack sizes up to 25 g and larger.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-003-04120 Supplier Medchemexpress LLC Supplier No. 25GHYW015782
4-Ethylresorcinol is a research-grade aromatic diol used in biochemical and cosmetic research to study pigmentation and tyrosinase activity. It has reported antioxidant activity and can inhibit melanin synthesis, making it useful for depigmentation studies. The compound is supplied as a high-purity solid and is provided with supporting documentation for research use.
  • Derivative of resorcinol that serves as a substrate for tyrosinase.
  • Reported antioxidant properties and inhibition of melanin synthesis.
  • High purity (99.71%) suitable for analytical and research assays.
  • Supplied as a solid powder with available pack sizes up to 25 g and larger.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet.