Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethyl-2,3-dioxo-1-piperazinecarbonylchloride, in Paraffin Oil (Tech.), 80%, Thermo Scientific™

Catalog No. AC259780500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC259780500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC259780500 Supplier Thermo Scientific Chemicals Supplier No. 259780500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59703-00-3
Molecular Formula C7H9ClN2O3
Molecular Weight (g/mol) 204.61
MDL Number MFCD00067054
InChI Key SXVBQOZRZIUHKU-UHFFFAOYSA-N
Synonym 4-ethyl-2,3-dioxo-1-piperazinecarbonyl chloride, 4-ethyl-2,3-dioxo-1-piperazinecarbonylchloride, 4-ethyl-2,3-dioxo-1-piperazine carbonyl chloride, 4-ethyl-2,3-dioxo-piperazine carbonyl chloride, 1-chlorocarbonyl-4-ethyl-2,3-dioxopiperazine, 4-ethyl-2,3-dioxo-piperazine-1-carbonyl chloride, 1-chlorocarbonyl-4-ethylpiperazine-2,3-dione, 2,3-dioxo-4-ethylpiperazine-1-carbonyl chloride, 4-ethylpiperazin-2,3-dione-1-ylcarbonyl chloride, 1-piperazinecarbonyl chloride, 4-ethyl-2,3-dioxo
PubChem CID 108813
IUPAC Name 4-ethyl-2,3-dioxopiperazine-1-carbonyl chloride
SMILES CCN1CCN(C(Cl)=O)C(=O)C1=O

Specifications

CAS Min % 75.0
CAS Max % 100.0
Color Tan to White
Infrared Spectrum Authentic
Quantity 50 g
Formula Weight 204.61
Percent Purity ≥75% (calculated on the standard color of a 1 w/v% min. soln. in methanol:%gt; 60% transmittance at 400nm)
Grade Technical
Physical Form Powder
Chemical Name or Material 4-Ethyl-2, 3-dioxo-1-piperazinecarbonylchloride, tech., 80%
Hazard Category MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.