Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylaniline, 99+%

Catalog No. AC118065000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118065000 500 g Glass bottle
AC118060050 5 g Glass bottle
AC118061000 100 g Glass bottle
AC118060025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC118065000 Supplier Thermo Scientific Chemicals Supplier No. 118065000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 589-16-2 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-16-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007921
InChI Key HRXZRAXKKNUKRF-UHFFFAOYSA-N
Synonym p-ethylaniline, benzenamine, 4-ethyl, 4-ethylphenylamine, aniline, 4-ethyl, p-ethylaminobenzene, 1-amino-4-ethylbenzene, 4-aminoethylbenzene, aniline, p-ethyl, unii-mhx72w1zqv, ccris 5060
PubChem CID 11504
IUPAC Name 4-ethylaniline
SMILES CCC1=CC=C(N)C=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -5°C
Color Brown-Red to Yellow
Density 0.9700g/mL
Boiling Point 216°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.5532 to 1.5552
Linear Formula C2H5C6H4NH2
Quantity 500 g
Beilstein 12, 1090
Specific Gravity 0.97
Solubility Information Solubility in water: immiscible
Formula Weight 121.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Ethylaniline, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-637-0
RTECSNumber BX9900000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.