Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylbenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
4748-78-1
C9H10O
134.18
MFCD00006956
QNGNSVIICDLXHT-UHFFFAOYSA-N
p-ethylbenzaldehyde,benzaldehyde, 4-ethyl,4-ethyl benzaldehyde,4-ethyl-benzaldehyde,ebal,ethyl benzaldehyde,ethyl-benzaldehyde,unii-289ppw3sg4,benzaldehyde,4-ethyl,ethylbenzaldehyde, p
20861
4-ethylbenzaldehyde
CCC1=CC=C(C=C1)C=O
4748-78-1
C9H10O
134.18
MFCD00006956
QNGNSVIICDLXHT-UHFFFAOYSA-N
p-ethylbenzaldehyde,benzaldehyde, 4-ethyl,4-ethyl benzaldehyde,4-ethyl-benzaldehyde,ebal,ethyl benzaldehyde,ethyl-benzaldehyde,unii-289ppw3sg4,benzaldehyde,4-ethyl,ethylbenzaldehyde, p
20861
4-ethylbenzaldehyde
CCC1=CC=C(C=C1)C=O
Boiling Point 221.0°C
Refractive Index 1.5370 to 1.5410
Packaging Glass bottle
Quantity 100g
Flash Point 92°C
Color Colorless to Yellow
Density 0.9700g/mL
Specific Gravity 0.97
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CH2C6H4CHO
Formula Weight 134.18
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Ethylbenzaldehyde
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-268-8