Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Ethylbenzoyl chloride

Catalog No. 2835095G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-350-95G 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-350-95G Supplier Sigma Aldrich Supplier No. 2835095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Ethylbenzoyl chloride

Specifications

CAS 16331-45-6
Density 1.147 g/mL (at 25°C (literature))
Boiling Point 235°C to 236°C (lit.)
Molecular Formula C9H9ClO
Refractive Index n20/D 1.5479 (literature)
Linear Formula C2H5C6H4COCl
MDL Number MFCD00000697
Quantity 50 mg
Molecular Weight (g/mol) 168.62
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.