Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylbenzoyl chloride, 97%

Catalog No. AC305730250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305730250 25 g Glass bottle
AC305730050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC305730250 Supplier Thermo Scientific Chemicals Supplier No. 305730250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16331-45-6 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16331-45-6
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000697
InChI Key AVTLLLZVYYPGFX-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-ethyl, p-ethylbenzoyl chloride, 4-ethylbenzoylchloride, 4-ethylbenzene-1-carbonyl chloride, 4-ethyl-benzoyl chloride, acmc-20amiy, pubchem9725, 4-ethyl benzoyl choride, 4-ethyl benzoyl chloride, benzoyl chloride,4-ethyl
PubChem CID 85368
IUPAC Name 4-ethylbenzoyl chloride
SMILES CCC1=CC=C(C=C1)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1400g/mL
Boiling Point 235°C to 236°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5465 to 1.5485
Linear Formula C2H5C6H4COCl
Quantity 25 g
Specific Gravity 1.14
Formula Weight 168.62
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Ethylbenzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-404-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.