Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-ethylcatechol | 1124-39-6 | MFCD00015847 | 99.5% | 138.17 g·mol⁻¹ | C8H10O2 | 10 G
SDP

Catalog No. 5000282318
Encompass_Preferred

4-Ethylcatechol is a dihydroxylated aromatic metabolite of 4-ethylphenol reported to induce oxidative DNA damage. It is supplied as a solid, research-grade chemical with defined purity, solubility, and storage recommendations suitable for laboratory use. Not for human or clinical use.

  • Purity: 99.5%
  • CAS number: 1124-39-6
  • Molecular formula: C8H10O2
  • Molecular weight: 138.17 g·mol⁻¹
  • Appearance: white to light brown solid
  • Solubility: ≥100 mg/mL in DMSO
  • Storage: 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month)
  • Intended use: research use only

Catalog No. 50-002-82318 Supplier Medchemexpress LLC Supplier No. HYW26575710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Ethylcatechol is a dihydroxylated aromatic metabolite of 4-ethylphenol reported to induce oxidative DNA damage. It is supplied as a solid, research-grade chemical with defined purity, solubility, and storage recommendations suitable for laboratory use. Not for human or clinical use.

  • Purity: 99.5%
  • CAS number: 1124-39-6
  • Molecular formula: C8H10O2
  • Molecular weight: 138.17 g·mol⁻¹
  • Appearance: white to light brown solid
  • Solubility: ≥100 mg/mL in DMSO
  • Storage: 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month)
  • Intended use: research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.