Learn More
Medchemexpress LLC 4-ethylcatechol | 1124-39-6 | MFCD00015847 | 99.5% | 138.17 g/mol | C8H10O2 | 5 G
SDP
Catalog No. 5000281835
Encompass_Preferred
4-Ethylcatechol is a research-grade ring-dihydroxylated metabolite of 4-ethylphenol reported to induce oxidative DNA damage. It is supplied as a high-purity solid for in vitro biochemical and cellular studies, metabolite research, and mechanistic assays.
  • High purity 99.5%.
  • Supplied as a solid suitable for storage under inert atmosphere.
  • Readily soluble in DMSO (≥100 mg/mL).
  • Molecular formula C8H10O2; molecular weight 138.17 g/mol.
  • Available in small research pack sizes, including a 5 G quantity.
Catalog No. 50-002-81835 Supplier Medchemexpress LLC Supplier No. HYW2657575G
4-Ethylcatechol is a research-grade ring-dihydroxylated metabolite of 4-ethylphenol reported to induce oxidative DNA damage. It is supplied as a high-purity solid for in vitro biochemical and cellular studies, metabolite research, and mechanistic assays.
  • High purity 99.5%.
  • Supplied as a solid suitable for storage under inert atmosphere.
  • Readily soluble in DMSO (≥100 mg/mL).
  • Molecular formula C8H10O2; molecular weight 138.17 g/mol.
  • Available in small research pack sizes, including a 5 G quantity.