Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-ethylcatechol | 1124-39-6 | MFCD00015847 | 99.5% | 138.17 g/mol | C8H10O2 | 500 MG
SDP

Supplier:  Medchemexpress LLC HYW265757500MG

Encompass_Preferred

4-Ethylcatechol is a catechol derivative and ring-dihydroxylated metabolite of 4-ethylphenol that induces oxidative DNA damage through redox cycling and reactive oxygen species generation; intended for research use only.

  • High purity: 99.5%.
  • Cas number: 1124-39-6.
  • Chemical formula: C8H10O2.
  • Molecular weight: 138.17 g/mol.
  • Soluble in DMSO at ≥100 mg/mL.
  • Storage: store under nitrogen; solid at 4°C; in solution -80°C up to 6 months or -20°C up to 1 month.
  • Applications: used as a DNA-damage agent in oxidative damage studies.

Catalog No. 50-002-80705


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.