Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. E09665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E09665G Supplier TCI America Supplier No. E09665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91328-77-7
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD01076323,MFCD01568857
InChI Key UNROFSAOTBVBBT-UHFFFAOYSA-N
Synonym trans-4-ethylcyclohexanecarboxylic acid, 4-ethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-ethyl, 4-trans-ethyl cyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-ethyl-, trans, 1s,4r-4-ethylcyclohexane-1-carboxylic acid, pubchem2491, acmc-209o2z, ksc352s1t, ksc352s2b
PubChem CID 1490288
IUPAC Name 4-ethylcyclohexane-1-carboxylic acid
SMILES CCC1CCC(CC1)C(O)=O

Specifications

Color Colorless
Quantity 5 g
Formula Weight 156.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Ethylcyclohexanecarboxylic Acid (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.