Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylcyclohexanol, 99%, mixture of cis- and trans isomers, Thermo Scientific™

Catalog No. AC181610250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC181610250 25 g
AC181611000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC181610250 Supplier Thermo Scientific Chemicals Supplier No. 181610250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4534-74-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00001450,MFCD00070694
InChI Key RVTKUJWGFBADIN-UHFFFAOYSA-N
Synonym 4-ethylcyclohexanol, trans-4-ethylcyclohexanol, cyclohexanol, 4-ethyl, cis-4-ethylcyclohexanol, cyclohexanol, 4-ethyl-, trans, 4-ethylcyclohexanol,c&t, 4-ethylcyclohexanol mixture of cis and trans, trans-4-ethyl cyclohexanol, ethyl cyclohexanol, cyclohexanol, 4-ethyl-, mixture of cis and trans
PubChem CID 78293
IUPAC Name 4-ethylcyclohexan-1-ol
SMILES CCC1CCC(O)CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.88g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
Linear Formula C2H5C6H10OH
Quantity 25 g
Beilstein 06, II, 26
Formula Weight 128.21
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 4-Ethylcyclohexanol, 99%
Show More Show Less
EINECSNumber 224-878-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.