Learn More
Sigma Aldrich Fine Chemicals Biosciences 4-Ethylmorpholine BioXtra, suitable for protein sequencing, >=99.5% (GC) | 100-74-3 | MFCD00006177 | 25ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 0449925ML
Encompass
4-Ethylmorpholine BioXtra, suitable for protein sequencing, >=99.5% (GC) | Purity: >=99.5% (GC) | Mol Wt: 115.17 | 100-74-3 | MFCD00006177 | 25ML
Catalog No. 50-188-3353