Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylphenetole 97.0+%, TCI America™
SDP

Catalog No. E043525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E043525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E043525ML Supplier TCI America Supplier No. E043525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1585-06-4
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00191432
InChI Key BQBROHBMIBOPFU-UHFFFAOYSA-N
Synonym 1-Ethoxy-4-ethylbenzene
PubChem CID 74107
IUPAC Name 1-ethoxy-4-ethylbenzene
SMILES CCC1=CC=C(C=C1)OCC

Specifications

Color Yellow
Boiling Point 205°C
Quantity 25 mL
Formula Weight 150.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylphenetole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.