Learn More
Apexbio Technology LLC 4-Ethylphenol 123-07-9 100mg
SDP
Catalog No. 5000090829
Encompass_Preferred
4-Ethylphenol (CAS 123-07-9) is a volatile phenolic compound predominantly produced through the microbial decarboxylation and reduction of p-coumaric acid during fermentation and other metabolic processes It is frequently utilized to investigate the microbial enzymatic pathways underlying aromatic compound metabolism In biomedical research 4-ethylphenol serves as a model molecule for studying the interplay between microbial metabolites and host metabolic networks as well as for the identification of microbial biomarkers in health and disease Its applications extend to experiments addressing the dynamics of microbial communities and host-microbe interactions
Catalog No. 50-000-90829 Supplier Apexbio Technology LLC Supplier No. C7126100
4-Ethylphenol (CAS 123-07-9) is a volatile phenolic compound predominantly produced through the microbial decarboxylation and reduction of p-coumaric acid during fermentation and other metabolic processes It is frequently utilized to investigate the microbial enzymatic pathways underlying aromatic compound metabolism In biomedical research 4-ethylphenol serves as a model molecule for studying the interplay between microbial metabolites and host metabolic networks as well as for the identification of microbial biomarkers in health and disease Its applications extend to experiments addressing the dynamics of microbial communities and host-microbe interactions