Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Ethylphenol | 123-07-9 | 99.99% | 122.17 | 1 ML
SDP

Catalog No. 5000454844
Encompass_Preferred

4-Ethylphenol is an endogenous metabolite and volatile phenolic compound. It is associated with off-odour in wine and can be synthesized by intestinal flora, where it is converted to 4-ethylphenyl sulfate. This compound causes DNA damage and induces 8-oxodG formation, and it also reduces oligodendrocyte maturation and myelination, increasing anxiety-like behavior in mouse models.

  • Purity: 99.99%
  • CAS number: 123-07-9
  • Molecular weight: 122.17
  • Formula: C8H10O
  • Appearance: Solid, white to light yellow
  • Structure classification: Phenols, Monophenols
  • Solubility: DMSO (60 mg/mL), H2O (2.63 mg/mL)
  • Applications: For research use only
  • Biological activity includes DNA damage, reduced oligodendrocyte maturation, and increased anxiety-like behavior

Catalog No. 50-004-54844 Supplier Medchemexpress LLC Supplier No. HYW01283610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Ethylphenol is an endogenous metabolite and volatile phenolic compound. It is associated with off-odour in wine and can be synthesized by intestinal flora, where it is converted to 4-ethylphenyl sulfate. This compound causes DNA damage and induces 8-oxodG formation, and it also reduces oligodendrocyte maturation and myelination, increasing anxiety-like behavior in mouse models.

  • Purity: 99.99%
  • CAS number: 123-07-9
  • Molecular weight: 122.17
  • Formula: C8H10O
  • Appearance: Solid, white to light yellow
  • Structure classification: Phenols, Monophenols
  • Solubility: DMSO (60 mg/mL), H2O (2.63 mg/mL)
  • Applications: For research use only
  • Biological activity includes DNA damage, reduced oligodendrocyte maturation, and increased anxiety-like behavior

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.