Learn More
Medchemexpress LLC 4-Ethylphenol | 123-07-9 | 99.99% | 122.17 | 100 G
SDP
Catalog No. 5000454939
Encompass_Preferred
4-Ethylphenol is a volatile phenolic compound commonly associated with off-odour in wine. This phenolic compound can be synthesized by intestinal flora and is converted to 4-ethylphenyl sulfate by *Lactobacillus plantarum*.
  • Causes DNA damage and induces 8-oxodG formation in calf thymus DNA in the presence of Cu (II)
  • Soluble in DMSO (60 mg/mL), H2O (2.63 mg/mL)
  • Available as a white to light yellow solid
  • Endogenous metabolite
Catalog No. 50-004-54939 Supplier Medchemexpress LLC Supplier No. HYW012836100G
4-Ethylphenol is a volatile phenolic compound commonly associated with off-odour in wine. This phenolic compound can be synthesized by intestinal flora and is converted to 4-ethylphenyl sulfate by *Lactobacillus plantarum*.
  • Causes DNA damage and induces 8-oxodG formation in calf thymus DNA in the presence of Cu (II)
  • Soluble in DMSO (60 mg/mL), H2O (2.63 mg/mL)
  • Available as a white to light yellow solid
  • Endogenous metabolite