Learn More
Medchemexpress LLC 4-Ethylphenol | 123-07-9 | MFCD00002393 | 98% | 122.16 g/mol | 500 G
SDP
Catalog No. 5000454882
Encompass_Preferred
4-Ethylphenol is a volatile phenolic compound associated with off-odour in wine. This compound can be synthesized by intestinal flora and is converted to 4-ethylphenyl sulfate by *Lactobacillus plantarum*.
  • Volatile phenolic compound
  • Synthesized by intestinal flora
  • Endogenous metabolite
  • Associated with neurological disease research
  • Classified as a monophenol
Catalog No. 50-004-54882 Supplier Medchemexpress LLC Supplier No. HYW012836500G
4-Ethylphenol is a volatile phenolic compound associated with off-odour in wine. This compound can be synthesized by intestinal flora and is converted to 4-ethylphenyl sulfate by *Lactobacillus plantarum*.
  • Volatile phenolic compound
  • Synthesized by intestinal flora
  • Endogenous metabolite
  • Associated with neurological disease research
  • Classified as a monophenol