Learn More
Selleck Chemical LLC 4-Ethylphenol
SDP
Catalog No. 5000003273
Encompass_Preferred
4-Ethylphenol (P-Ethylphenol 4-Hydroxyphenylethane) is a phenolic compound produced in wine and beer by the spoilage yeast Brettanomyces
Catalog No. 50-000-03273 Supplier Selleck Chemical LLC Supplier No. S630825MG
4-Ethylphenol (P-Ethylphenol 4-Hydroxyphenylethane) is a phenolic compound produced in wine and beer by the spoilage yeast Brettanomyces