Learn More
Medchemexpress LLC 4-Ethylphenol-d10 | 352431-18-6 | 97.9% | 132.23 | 25 MG
SDP
Catalog No. 5000366816
Encompass_Preferred
4-Ethylphenol-d10 is the deuterium-labeled version of 4-Ethylphenol. 4-Ethylphenol is a volatile phenolic compound linked to off-odors in wine and is a phenolic compound synthesized by intestinal flora. It is converted to 4-ethylphenyl sulfate by *Lactobacillus plantarum*.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-66816 Supplier Medchemexpress LLC Supplier No. 25MGHYW012836S1
4-Ethylphenol-d10 is the deuterium-labeled version of 4-Ethylphenol. 4-Ethylphenol is a volatile phenolic compound linked to off-odors in wine and is a phenolic compound synthesized by intestinal flora. It is converted to 4-ethylphenyl sulfate by *Lactobacillus plantarum*.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.