Learn More
Medchemexpress LLC 4-Ethylphenol-d10 | 352431-18-6 | 97.91% | 132.23 | 5 MG
SDP
Catalog No. 5000454804
Encompass_Preferred
4-Ethylphenol-d10 is the deuterium-labeled version of 4-Ethylphenol. 4-Ethylphenol is a volatile phenolic compound linked to off-odour in wine and can be synthesized by intestinal flora. It is converted to 4-ethylphenyl sulfate by *Lactobacillus plantarum*.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes (hydrogen, carbon, and other elements) into drug molecules can serve as tracers for quantitation during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54804 Supplier Medchemexpress LLC Supplier No. HYW012836S15MG
4-Ethylphenol-d10 is the deuterium-labeled version of 4-Ethylphenol. 4-Ethylphenol is a volatile phenolic compound linked to off-odour in wine and can be synthesized by intestinal flora. It is converted to 4-ethylphenyl sulfate by *Lactobacillus plantarum*.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes (hydrogen, carbon, and other elements) into drug molecules can serve as tracers for quantitation during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.