Learn More
Apexbio Technology LLC 4-ethylphenyl sulfate(Synonyms: 4-Ethylphenyl sulfate, 4-ethylphenyl hydrogen sulfate, p-Ethylphenyl sulfate), 25mg.
SDP
Catalog No. 5000060219
Encompass_Preferred
4-Ethylphenyl sulfate is a microbiota-derived metabolite structurally related to p-cresol (4-methylphenol) and classified as a uremic toxin Elevated serum concentrations have been detected in chronic renal failure patients suggesting potential utility as a biomarker for renal function In murine models of autism induced by maternal immune activation (MIA) serum levels of 4-ethylphenyl sulfate significantly increased Administration to healthy mice induced anxiety-like behaviors and amplified startle sensitivity without affecting pre-pulse inhibition indicating involvement in behavioral and neurological modulation This compound serves as a promising research tool for studying gut microbiota-brain interactions in autism-spectrum disorders and renal dysfunction
Catalog No. 50-000-60219 Supplier Apexbio Technology LLC Supplier No. B605125
4-Ethylphenyl sulfate is a microbiota-derived metabolite structurally related to p-cresol (4-methylphenol) and classified as a uremic toxin Elevated serum concentrations have been detected in chronic renal failure patients suggesting potential utility as a biomarker for renal function In murine models of autism induced by maternal immune activation (MIA) serum levels of 4-ethylphenyl sulfate significantly increased Administration to healthy mice induced anxiety-like behaviors and amplified startle sensitivity without affecting pre-pulse inhibition indicating involvement in behavioral and neurological modulation This compound serves as a promising research tool for studying gut microbiota-brain interactions in autism-spectrum disorders and renal dysfunction