Learn More
Medchemexpress LLC 4-ethylresorcinol | 2896-60-8 | MFCD00002283 | 99.7% | 138.16 g/mol | C8H10O2 | 50 G
SDP
Catalog No. 5000300324
Encompass_Preferred
4-Ethylresorcinol (CAS 2896-60-8) is a resorcinol derivative used in research for its tyrosinase-inhibiting, antioxidant, and skin-depigmenting properties. It is supplied as a high-purity solid and as a ready-to-use DMSO solution for laboratory studies of melanin synthesis, cosmetic active evaluation, and related biochemical assays.
  • High purity (≈99.7%) for reliable experimental results.
  • Inhibits tyrosinase and helps reduce melanin synthesis.
  • Exhibits antioxidant activity relevant to skin research.
  • Available as solid powder and 10 mM solution in DMSO.
  • Suitable for cosmetic and biochemical research applications.
Catalog No. 50-003-00324 Supplier Medchemexpress LLC Supplier No. HYW01578250G
4-Ethylresorcinol (CAS 2896-60-8) is a resorcinol derivative used in research for its tyrosinase-inhibiting, antioxidant, and skin-depigmenting properties. It is supplied as a high-purity solid and as a ready-to-use DMSO solution for laboratory studies of melanin synthesis, cosmetic active evaluation, and related biochemical assays.
  • High purity (≈99.7%) for reliable experimental results.
  • Inhibits tyrosinase and helps reduce melanin synthesis.
  • Exhibits antioxidant activity relevant to skin research.
  • Available as solid powder and 10 mM solution in DMSO.
  • Suitable for cosmetic and biochemical research applications.