Learn More
Medchemexpress LLC 4-ethylresorcinol | 2896-60-8 | MFCD00002283 | 99.7% | 138.17 | C8H10O2 | 1 ML
SDP
Catalog No. 5000300755
Encompass_Preferred
4-Ethylresorcinol is a phenolic small molecule used in biochemical research to investigate tyrosinase activity, melanin synthesis, and antioxidant properties. It is supplied as a ready-to-use solution and in solid form for assay preparation and formulation.
  • Ready-to-use 10 mM solution in DMSO for in vitro assays.
  • High reported purity, suitable for analytical and biological studies.
  • Also available as solid quantities for custom dilution and formulation.
  • Phenolic structure useful for studies of pigmentation and antioxidant activity.
  • Documented chemical identifiers and supporting datasheet and COA available.
Catalog No. 50-003-00755 Supplier Medchemexpress LLC Supplier No. HYW01578210MM/1ML
4-Ethylresorcinol is a phenolic small molecule used in biochemical research to investigate tyrosinase activity, melanin synthesis, and antioxidant properties. It is supplied as a ready-to-use solution and in solid form for assay preparation and formulation.
  • Ready-to-use 10 mM solution in DMSO for in vitro assays.
  • High reported purity, suitable for analytical and biological studies.
  • Also available as solid quantities for custom dilution and formulation.
  • Phenolic structure useful for studies of pigmentation and antioxidant activity.
  • Documented chemical identifiers and supporting datasheet and COA available.