Learn More
Medchemexpress LLC 4-ethylresorcinol | 2896-60-8 | MFCD00002283 | 99.7% | 138.17 | C8H10O2 | 10 G
SDP
Catalog No. 5000300061
Encompass_Preferred
4-Ethylresorcinol is a resorcinol derivative used in research as a tyrosinase substrate with reported anti-hyperpigmentation and antioxidant activity. It reduces melanin synthesis by inhibiting TRP-2 expression, lowering cAMP levels, and suppressing PKA and tyrosinase activity in melanocyte models; it also inhibits lipid peroxidation in liver microsomes.
  • High purity: 99.71% suitable for analytical and biochemical assays.
  • Functionally active in melanogenesis and antioxidation studies.
  • Solid small molecule with defined molecular formula and weight for accurate dosing.
  • Compatible with tyrosinase activity and cell-based pigmentation assays.
  • Available in multiple pack sizes to support pilot and scale studies.
Catalog No. 50-003-00061 Supplier Medchemexpress LLC Supplier No. HYW01578210G
4-Ethylresorcinol is a resorcinol derivative used in research as a tyrosinase substrate with reported anti-hyperpigmentation and antioxidant activity. It reduces melanin synthesis by inhibiting TRP-2 expression, lowering cAMP levels, and suppressing PKA and tyrosinase activity in melanocyte models; it also inhibits lipid peroxidation in liver microsomes.
  • High purity: 99.71% suitable for analytical and biochemical assays.
  • Functionally active in melanogenesis and antioxidation studies.
  • Solid small molecule with defined molecular formula and weight for accurate dosing.
  • Compatible with tyrosinase activity and cell-based pigmentation assays.
  • Available in multiple pack sizes to support pilot and scale studies.