Learn More
Medchemexpress LLC 4-ethylresorcinol | 2896-60-8 | MFCD00002283 | 99.7% | 138.17 g/mol | C8H10O2 | 1 G
SDP
Catalog No. 5000300076
Encompass_Preferred
4-Ethylresorcinol is a small-molecule research reagent used as a depigmenting agent and tyrosinase modulator. It is supplied as an off-white to yellow solid with high stated purity and is employed in biochemical and cell-based studies that probe melanin synthesis, enzyme inhibition, and related pathways.
  • High purity suitable for research applications.
  • Off-white to yellow solid for ease of handling.
  • Used as a depigmenting agent and tyrosinase inhibitor in assays.
  • Available as ready-to-use solution in DMSO or as a solid for custom preparation.
  • Offered in multiple pack sizes to support small- and large-scale needs.
  • Molecular weight 138.17 g/mol and formula C8H10O2 for reference.
Catalog No. 50-003-00076 Supplier Medchemexpress LLC Supplier No. HYW0157821G
4-Ethylresorcinol is a small-molecule research reagent used as a depigmenting agent and tyrosinase modulator. It is supplied as an off-white to yellow solid with high stated purity and is employed in biochemical and cell-based studies that probe melanin synthesis, enzyme inhibition, and related pathways.
  • High purity suitable for research applications.
  • Off-white to yellow solid for ease of handling.
  • Used as a depigmenting agent and tyrosinase inhibitor in assays.
  • Available as ready-to-use solution in DMSO or as a solid for custom preparation.
  • Offered in multiple pack sizes to support small- and large-scale needs.
  • Molecular weight 138.17 g/mol and formula C8H10O2 for reference.