Learn More
Medchemexpress LLC 4-ethylresorcinol | 2896-60-8 | MFCD00002283 | 99.7% | 138.17 g/mol | C8H10O2 | 5 G
SDP
Catalog No. 5000300325
Encompass_Preferred
4-ethylresorcinol is a high-purity resorcinol derivative used as a tyrosinase substrate and depigmenting agent in research. It is supplied as a solid suitable for in vitro assays, analytical standards, and method development.
  • High purity: 99.71%.
  • Chemical formula: C8H10O2; molecular weight: 138.17 g/mol.
  • Suitable as a tyrosinase substrate and for depigmentation studies.
  • Available in multiple sizes, including milligram to bulk quantities.
  • Documentation available: datasheet, COA, SDS, and spectral data.
Catalog No. 50-003-00325 Supplier Medchemexpress LLC Supplier No. HYW0157825G
4-ethylresorcinol is a high-purity resorcinol derivative used as a tyrosinase substrate and depigmenting agent in research. It is supplied as a solid suitable for in vitro assays, analytical standards, and method development.
  • High purity: 99.71%.
  • Chemical formula: C8H10O2; molecular weight: 138.17 g/mol.
  • Suitable as a tyrosinase substrate and for depigmentation studies.
  • Available in multiple sizes, including milligram to bulk quantities.
  • Documentation available: datasheet, COA, SDS, and spectral data.