Learn More
Medchemexpress LLC 4-Ethynylaniline | 14235-81-5 | 117.15 | 1 ML
SDP
Catalog No. 5000448880
Encompass_Preferred
4-Ethynylaniline is a biochemical reagent used in life science research. It serves as a click chemistry reagent, specifically containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Used as a biochemical reagent
  • Utilized in life science research
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with azide-containing molecules
Catalog No. 50-004-48880 Supplier Medchemexpress LLC Supplier No. HYW00466010MM/1ML
4-Ethynylaniline is a biochemical reagent used in life science research. It serves as a click chemistry reagent, specifically containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Used as a biochemical reagent
  • Utilized in life science research
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with azide-containing molecules