Learn More
Medchemexpress LLC 4-Ethynylaniline | 14235-81-5 | 117.15 | 10 G
Supplier: Medchemexpress LLC HYW00466010G
4-Ethynylaniline is a biochemical reagent used as a biological material or organic compound for life science-related research. It is a click chemistry reagent containing an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. Click chemistry is characterized by selective, modular, wide-ranging, and high-yield chemical reactions that facilitate rapid synthesis of new compounds through heteroatom linking (C-X-C).
- Contains an alkyne group
- Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
- Enables rapid synthesis of new compounds
- Used in bioorthogonal chemistry
- For research use only