Learn More
Medchemexpress LLC 4-Ethynylaniline | 14235-81-5 | 117.15 | 5 G
SDP
Catalog No. 5000448881
Encompass_Preferred
4-Ethynylaniline is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is a click chemistry reagent containing an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used in click chemistry
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Biochemical assay reagent
Catalog No. 50-004-48881 Supplier Medchemexpress LLC Supplier No. HYW0046605G
4-Ethynylaniline is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is a click chemistry reagent containing an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used in click chemistry
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Biochemical assay reagent