Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethynyltriphenylamine 98.0+%, TCI America™
SDP

Catalog No. E08941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08941G Supplier TCI America Supplier No. E08941G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 205877-26-5
Molecular Formula C20H15N
Molecular Weight (g/mol) 269.347
MDL Number MFCD09030769
InChI Key QDYFCLVSLUZDHB-UHFFFAOYSA-N
Synonym 4-Diphenylaminophenylacetylene
PubChem CID 20612855
IUPAC Name 4-ethynyl-N,N-diphenylaniline
SMILES C#CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 111°C
Color Yellow
Quantity 1 g
Formula Weight 269.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethynyltriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.