Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Feruloylquinic acid | 2613-86-7 | 5 MG
SDP

Catalog No. 5000418073
Encompass_Preferred

4-Feruloylquinic acid, with a purity of 99.41%, is a chemical compound that may serve as a potential biomarker for food products. This white to off-white solid has a molecular weight of 368.34 and the chemical formula C17H20O9. It is intended for research use only.

  • Potential biomarker for food products
  • High purity of 99.41%
  • Molecular formula: C17H20O9
  • Molecular weight: 368.34
  • Appearance: White to off-white solid
  • Classified under Phenylpropanoids, Simple Phenylpropanols, Phenols, and Monophenols
  • Initial source from plants
  • In vitro solubility in DMSO: 50 mg/mL (135.74 mM)
  • Various in vivo dissolution protocols available

Catalog No. 50-004-18073 Supplier Medchemexpress LLC Supplier No. HYN65985MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Feruloylquinic acid, with a purity of 99.41%, is a chemical compound that may serve as a potential biomarker for food products. This white to off-white solid has a molecular weight of 368.34 and the chemical formula C17H20O9. It is intended for research use only.

  • Potential biomarker for food products
  • High purity of 99.41%
  • Molecular formula: C17H20O9
  • Molecular weight: 368.34
  • Appearance: White to off-white solid
  • Classified under Phenylpropanoids, Simple Phenylpropanols, Phenols, and Monophenols
  • Initial source from plants
  • In vitro solubility in DMSO: 50 mg/mL (135.74 mM)
  • Various in vivo dissolution protocols available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.