Learn More
Ace Glass, Inc. Column extender, 4" ID x 24" , 4" O-ring grooved flat flange top, 4" ground flat flange bottom
SDP
Catalog No. 50978540
Encompass_Preferred
4 I.D. x 24 column extender with size 4 o-ring grooved flat flange top and size 4 ground flat flange bottom. Column extender can be mated to column with ACE 6517 series clamps.
Catalog No. 50-978-540 Supplier Ace Glass, Inc. Supplier No. 586274
4 I.D. x 24 column extender with size 4 o-ring grooved flat flange top and size 4 ground flat flange bottom. Column extender can be mated to column with ACE 6517 series clamps.