Learn More
Julabo USA Inc Pair, Quad distributing adapter M16x1f to 4x M16x1m, for FC chillers
SDP
Catalog No. 507534227
Encompass_Preferred
Bath Accessories, Julabo, Quad distributing adapter (2 pieces), M16x1 female to 4x M16x1 male fittings, with barbed fittings for tubing 8 mm or 12 mm / 1/2″ ID. For use with FC recirculating coolers.
Catalog No. 50-753-4227 Supplier Julabo USA Inc Supplier No. 8970474
Bath Accessories, Julabo, Quad distributing adapter (2 pieces), M16x1 female to 4x M16x1 male fittings, with barbed fittings for tubing 8 mm or 12 mm / 1/2″ ID. For use with FC recirculating coolers.