Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-1,3-phenylenediamine, 95%

Catalog No. AAH6147606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6147606 5 g
AAH6147603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6147606 Supplier Thermo Scientific Chemicals Supplier No. H6147606

missing translation for 'validMainframeMsg'

CAS: 6264-67-1 | C6H7FN2 | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6264-67-1
Molecular Formula C6H7FN2
Molecular Weight (g/mol) 126.13
MDL Number MFCD00077494
InChI Key QNDFYLBDUWCFJO-UHFFFAOYSA-N
PubChem CID 80435
IUPAC Name 4-fluorobenzene-1,3-diamine
SMILES NC1=CC=C(F)C(N)=C1

Specifications

Melting Point 34°C to 37°C
Color Black
Boiling Point 152°C to 153°C
Flash Point 105°C (221°F)
Quantity 5 g
Synonym 2,4-diaminofluorobenzene, 4-fluoro-1,3-benzenediamine, 4-fluoro-1,3-phenylenediamine, 1,3-diamino-4-fluorobenzene, 1,3-benzenediamine, 4-fluoro, 4-fluoro-1,3-diaminobenzene, 1,3-benzenediamine,4-fluoro, pubchem2538, acmc-209n6n, ksc615i4l
Formula Weight 126.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Fluoro-1,3-phenylenediamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-426-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.