Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-1,3-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. F07945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F07945G Supplier TCI America Supplier No. F07945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6264-67-1
Molecular Formula C6H7FN2
Molecular Weight (g/mol) 126.13
MDL Number MFCD00077494
InChI Key QNDFYLBDUWCFJO-UHFFFAOYSA-N
Synonym 2,4-diaminofluorobenzene, 4-fluoro-1,3-benzenediamine, 4-fluoro-1,3-phenylenediamine, 1,3-diamino-4-fluorobenzene, 1,3-benzenediamine, 4-fluoro, 4-fluoro-1,3-diaminobenzene, 1,3-benzenediamine,4-fluoro, pubchem2538, acmc-209n6n, ksc615i4l
PubChem CID 80435
IUPAC Name 4-fluorobenzene-1,3-diamine
SMILES NC1=CC=C(F)C(N)=C1

Specifications

Melting Point 36°C
Boiling Point 153°C
Quantity 5 g
Formula Weight 126.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Fluoro-1,3-phenylenediamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.