Learn More
Aobchem 4-Fluoro-2-iodopyridine | ≥95% | CAS 2385268-22-2 | C5H3FIN | 250mg
Catalog No. 5000903958
Encompass_Preferred
4-Fluoro-2-iodopyridine | ≥95% | CAS 2385268-22-2 | C5H3FIN | 250mg
Catalog No. 50-009-03958 Supplier Aobchem Supplier No. A27151250MG
4-Fluoro-2-iodopyridine | ≥95% | CAS 2385268-22-2 | C5H3FIN | 250mg