Learn More
Accela Chembio Inc 4-fluoro-2-methylbenzoic Acid | 25g | 321-21-1 | MFCD04115880 | 97+% | Shelf Life: 1800 Days | Regular
Catalog No. 50354461
Encompass_Preferred
4-fluoro-2-methylbenzoic Acid | 25g | 321-21-1 | MFCD04115880 | 97+% | Shelf Life: 1800 Days | Regular
Catalog No. 50-354-461 Supplier Accela Chembio Inc Supplier No. SY00613625G
Add to Cart
Add to Cart
4-fluoro-2-methylbenzoic Acid | 25g | 321-21-1 | MFCD04115880 | 97+% | Shelf Life: 1800 Days | Regular