Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methylbenzoic acid, 99%, Thermo Scientific™

Catalog No. AC399300010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399300010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399300010 Supplier Thermo Scientific Chemicals Supplier No. 399300010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 321-21-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD04115880
InChI Key KDXOONIQRUZGSY-UHFFFAOYSA-N
Synonym 4-fluoro-o-toluic acid, 2-methyl-4-fluorobenzoic acid, 4-fluoro-2-methylbenzoicacid, 4-fluoro-2-methyl-benzoic acid, benzoic acid, 4-fluoro-2-methyl, pubchem1327, 2-carboxy-5-fluorotoluene, acmc-209hr6, 4-fluoro-2-methyl benzoicacid, rarechem al be 0506
PubChem CID 3860639
IUPAC Name 4-fluoro-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)F)C(=O)O

Specifications

Melting Point 168.0°C to 172.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H3(CH3)COOH
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 154.14
Percent Purity 99%
Chemical Name or Material 4-Fluoro-2-methylbenzoic acid
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.