Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methylbenzonitrile, 97%, Thermo Scientific™

Catalog No. AC399930050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399930050 5 g Glass bottle
AC399930010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399930050 Supplier Thermo Scientific Chemicals Supplier No. 399930050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 147754-12-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD03095106
InChI Key BJBXUIUJKPOZLV-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorobenzonitrile, 4-fluoro-2-methyl-benzonitrile, benzonitrile, 4-fluoro-2-methyl, 2-cyano-5-fluorotoluene, 4-fluoro-2-methylbenzenecarbonitrile, pubchem1554, intermediates-zcf02129, methyl 4-fluorobenzonitrile, 4-fuoro-2-methylbenzonitrile, ksc174s6l
PubChem CID 3826055
IUPAC Name 4-fluoro-2-methylbenzonitrile
SMILES CC1=C(C=CC(=C1)F)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 70.0°C to 74.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(CH3)CN
Quantity 5 g
Formula Weight 135.14
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylbenzonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.