Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F10525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1052-5G 5 g
F10521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1052-5G Supplier TCI America Supplier No. F10525G
Add to Cart
Add to Cart

Specifications

CAS 182344-16-7
Melting Point 186°C
Color White
Molecular Formula C7H5BF4O2
MDL Number MFCD08062379
Quantity 5 g
Synonym 4-fluoro-2-trifluoromethyl phenyl boronic acid, 4-fluoro-2-trifluoromethyl benzeneboronic acid, 4-fluoro-2-trifluoromethyl phenylboronic acid, 2-borono-5-fluorobenzotrifluoride, boronic acid, 4-fluoro-2-trifluoromethyl phenyl
InChI Key SWUPLEAGZOKLNX-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)F)C(F)(F)F)(O)O
IUPAC Name [4-fluoro-2-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 207.919
PubChem CID 12109469
Formula Weight 207.92
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.