Learn More
Medchemexpress LLC 4-fluoro-3-[(4-hydroxy-1-piperidinyl)sulfonyl]-N-(3,4,5-trifluorophenyl)-benzamide | 1445790-55-5 | 99.9% | 432.39 g·mol⁻¹ | C18H16F4N2O4S | 10MG
SDP
Catalog No. 5000236583
Encompass_Preferred
NVR 3-778 is a first-in-class, orally bioavailable hepatitis B virus (HBV) capsid assembly modulator of the sulfamoylbenzamide class. It inhibits HBV capsid assembly and viral replication, shows pan-genotypic antiviral activity in preclinical models, and has advanced into clinical evaluation. Supplied as a high-purity research-grade compound for in vitro and in vivo HBV studies.
  • High purity (99.9%).
  • Orally bioavailable in preclinical models.
  • Pan-genotypic antiviral activity against HBV.
  • Mechanism: capsid assembly modulation.
  • Available in multiple small pack sizes for research flexibility.
  • Stable as powder at -20°C for up to 3 years.
Catalog No. 50-002-36583 Supplier Medchemexpress LLC Supplier No. HY12460010MG
NVR 3-778 is a first-in-class, orally bioavailable hepatitis B virus (HBV) capsid assembly modulator of the sulfamoylbenzamide class. It inhibits HBV capsid assembly and viral replication, shows pan-genotypic antiviral activity in preclinical models, and has advanced into clinical evaluation. Supplied as a high-purity research-grade compound for in vitro and in vivo HBV studies.
  • High purity (99.9%).
  • Orally bioavailable in preclinical models.
  • Pan-genotypic antiviral activity against HBV.
  • Mechanism: capsid assembly modulation.
  • Available in multiple small pack sizes for research flexibility.
  • Stable as powder at -20°C for up to 3 years.