Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-fluoro-3-(4-hydroxypiperidin-1-yl)sulfonyl-N-(3,4,5-trifluorophenyl)benzamide | 1445790-55-5 | 99.9% | 432.39 g·mol-1 | C18H16F4N2O4S | 5MG
SDP

Catalog No. 5000237452
Encompass_Preferred

NVR 3-778 is a research-stage hepatitis B virus (HBV) capsid assembly modulator (CAM) belonging to the sulfamoylbenzamide class. It is orally bioavailable and has demonstrated antiviral activity in preclinical and clinical studies; the compound is supplied as a solid powder or as a DMSO solution for use in in vitro and in vivo research.

  • Reported purity 99.88%.
  • Molecular formula C18H16F4N2O4S and molecular weight 432.39 g·mol-1.
  • CAS number 1445790-55-5.
  • Available in small research quantities and as a 10 mM solution in DMSO.
  • Used for mechanism-of-action, antiviral efficacy, and pharmacokinetic studies.

Catalog No. 50-002-37452 Supplier Medchemexpress LLC Supplier No. HY1246005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NVR 3-778 is a research-stage hepatitis B virus (HBV) capsid assembly modulator (CAM) belonging to the sulfamoylbenzamide class. It is orally bioavailable and has demonstrated antiviral activity in preclinical and clinical studies; the compound is supplied as a solid powder or as a DMSO solution for use in in vitro and in vivo research.

  • Reported purity 99.88%.
  • Molecular formula C18H16F4N2O4S and molecular weight 432.39 g·mol-1.
  • CAS number 1445790-55-5.
  • Available in small research quantities and as a 10 mM solution in DMSO.
  • Used for mechanism-of-action, antiviral efficacy, and pharmacokinetic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.