Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-fluoro-3-(4-hydroxypiperidin-1-yl)sulfonyl-N-(3,4,5-trifluorophenyl)benzamide | 1445790-55-5 | 99.9% | 432.39 g/mol | C18H16F4N2O4S | 10MM 1ML
SDP

Catalog No. 5000236104
Encompass_Preferred

NVR 3-778 is a small-molecule hepatitis B virus (HBV) capsid assembly modulator (CAM) of the sulfamoylbenzamide class with reported oral bioavailability and anti-HBV activity. It is supplied for research use as a high-purity solid and is commonly provided as 10 mM stock solutions in DMSO for in vitro assays.

  • First-in-class capsid assembly modulator for HBV research.
  • Orally bioavailable compound with reported antiviral activity.
  • High purity (≈99.9%) suitable for analytical and biological studies.
  • Molecular weight 432.39 g/mol; formula C18H16F4N2O4S.
  • Available as solid and as 10 mM DMSO solution for convenience in assays.

Catalog No. 50-002-36104 Supplier Medchemexpress LLC Supplier No. HY12460010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NVR 3-778 is a small-molecule hepatitis B virus (HBV) capsid assembly modulator (CAM) of the sulfamoylbenzamide class with reported oral bioavailability and anti-HBV activity. It is supplied for research use as a high-purity solid and is commonly provided as 10 mM stock solutions in DMSO for in vitro assays.

  • First-in-class capsid assembly modulator for HBV research.
  • Orally bioavailable compound with reported antiviral activity.
  • High purity (≈99.9%) suitable for analytical and biological studies.
  • Molecular weight 432.39 g/mol; formula C18H16F4N2O4S.
  • Available as solid and as 10 mM DMSO solution for convenience in assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.