Learn More
Medchemexpress LLC 4-fluoro-3-(4-hydroxypiperidin-1-yl)sulfonyl-N-(3,4,5-trifluorophenyl)benzamide | 1445790-55-5 | 99.9% | 432.39 g/mol | C18H16F4N2O4S | 50MG
SDP
Catalog No. 5000237099
Encompass_Preferred
NVR 3-778 is a first-in-class, orally bioavailable hepatitis B virus (HBV) capsid assembly modulator (CAM) of the sulfamoylbenzamide chemical class used in research to inhibit HBV replication and the generation of infectious viral particles. It has been characterized in preclinical and clinical studies and is supplied for research use with high reported purity and defined physicochemical properties.
  • First-in-class capsid assembly modulator with demonstrated antiviral activity.
  • Orally bioavailable small molecule suitable for preclinical assays.
  • Pan-genotypic inhibition of HBV replication in cell-based models.
  • Solid, white to off-white material with reported solubility in DMSO and ethanol.
  • High reported purity appropriate for research applications.
Catalog No. 50-002-37099 Supplier Medchemexpress LLC Supplier No. HY12460050MG
NVR 3-778 is a first-in-class, orally bioavailable hepatitis B virus (HBV) capsid assembly modulator (CAM) of the sulfamoylbenzamide chemical class used in research to inhibit HBV replication and the generation of infectious viral particles. It has been characterized in preclinical and clinical studies and is supplied for research use with high reported purity and defined physicochemical properties.
  • First-in-class capsid assembly modulator with demonstrated antiviral activity.
  • Orally bioavailable small molecule suitable for preclinical assays.
  • Pan-genotypic inhibition of HBV replication in cell-based models.
  • Solid, white to off-white material with reported solubility in DMSO and ethanol.
  • High reported purity appropriate for research applications.