Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F10615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1061-5G 5 g
F10611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1061-5G Supplier TCI America Supplier No. F10615G
Add to Cart
Add to Cart

Specifications

CAS 374538-01-9
Melting Point 220°C
Color White-Yellow
Molecular Formula C7H6BFO3
MDL Number MFCD02093074
Quantity 5 g
Synonym 4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809
InChI Key YABSTJQEBSKPCG-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)F)C=O)(O)O
IUPAC Name (4-fluoro-3-formylphenyl)boronic acid
Molecular Weight (g/mol) 167.93
PubChem CID 2778651
Formula Weight 167.93
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.