Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-hydroxybenzeneboronic acid, 98%, Thermo Scientific™

Catalog No. AAH52550MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52550MD 250 mg
AAH5255003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52550MD Supplier Thermo Scientific Chemicals Supplier No. H52550MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluoro-3-hydroxybenzeneboronic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-74-2
Molecular Formula C6H6BFO3
Molecular Weight (g/mol) 155.919
MDL Number MFCD08689483
InChI Key GNCXNRGBOBWFSO-UHFFFAOYSA-N
Synonym 4-fluoro-3-hydroxyphenyl boronic acid, 4-fluoro-3-hydroxybenzeneboronic acid, 4-fluoro-3-hydroxy-phenyl boronic acid, 5-borono-2-fluorophenol, acmc-209r9z, 4-fluoro-3-hydroxy-phenylboronic acid, 4-fluoranyl-3-oxidanyl-phenyl boronic acid, boronic acid, b-4-fluoro-3-hydroxyphenyl
PubChem CID 44717470
IUPAC Name (4-fluoro-3-hydroxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)O)(O)O

Specifications

Melting Point 264°C to 266°C
Quantity 250 mg
Formula Weight 155.92
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-hydroxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.