Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-methylbenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. F07915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07915G 5 g
F079125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07915G Supplier TCI America Supplier No. F07915G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135427-08-6
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.141
MDL Number MFCD01631431
InChI Key NRFKZFFVTGGEQF-UHFFFAOYSA-N
Synonym 4-fluoro-3-methyl benzaldehyde, benzaldehyde, 4-fluoro-3-methyl, 3-methyl-4-fluorobenzaldehyde, 4-fluoro-3-methyl-benzaldehyde, 4-fluoro-3-methylbenzaldehyde, pubchem1457, acmc-1chue, ksc494g7n, pharmabridge p-3415, 3-methyl-4-fluoro benzaldehyde
PubChem CID 2734874
IUPAC Name 4-fluoro-3-methylbenzaldehyde
SMILES CC1=C(C=CC(=C1)C=O)F

Specifications

Color Yellow
Boiling Point 208°C
Quantity 5 g
Formula Weight 138.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-methylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.